Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIAS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597924
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597924 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597924 Supplier Invitrogen™ Supplier No. PA597924
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LIAS Polyclonal Antibody for Western Blot, IHC (P), ELISA

LIAS belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis.The protein encoded by this gene belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.The protein encoded by this gene belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIAS
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Lias
Gene Accession No. O43766
Gene Alias 2900022L22Rik; 4933425M12Rik; 7a5ex; C77512; HAMAP-Rule:MF_03123}; HGCLAS; HUSSY-01; hypothetical protein MGC66080; LAS; LIAS; LIAS protein; LIP1; Lipoate synthase; lipoate synthase {ECO:0000255; lipoic acid synthase; lipoic acid synthase {ECO:0000255; lipoic acid synthetase; lipoic acid synthetase isoform 1 precursor; lipoyl synthase, mitochondrial; lipoyl synthase, mitochondrial {ECO:0000255; Lipoyl synthase, mitochondrial-like protein; lip-syn; lip-syn {ECO:0000255; LS; LS {ECO:0000255; MGC23245; mLip1; PDHLD; zgc:66080
Gene Symbols Lias
Host Species Rabbit
Immunogen Recombinant Human Lipoyl synthase, mitochondrial protein (28-372AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11019
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.