Learn More
Abcam Licarbazepine, 10,11-Dihydro-10-hydroxycarbamazepine, Solid, >99% Purity, 10 mg
Description
- Molecular weight: 254.28 Da
- Purity: >99%
- Physical form: Solid
- Activity: Active metabolite of oxcarbazepine; induces microsomal enzymes via sodium channel protein inhibition; potentiates minor GABAA receptor currents; exhibits anticonvulsant activity
- Application: Antiepileptic agent; active in vivo
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Sodium Channel Protein, GABAA Receptor |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Quantity | 10 mg |
| For Use With (Application) | Anticonvulsant activity |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.