Learn More
Mettler Toledo™ License EasyDirect Balance
Catalog No. 01915038 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Includes:
10 Licenses
3 Licenses
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
01-915-038 3 Licenses
01-915-039 10 Licenses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-915-038 Supplier Mettler Toledo™ Supplier No. 30539323
Add to Cart
Add to Cart
Collect weighing data from up to 10 advanced and standard level balances on one PC via Ethernet or RS232 interface. Easily review results, generate reports, and export data in various formats.
  • Collect weighing data for multiple advanced and standard level balances on one PC
  • Software helps to minimize transcription errors, save time, and securely store data
  • Visualize results in charts to asses target/tolerance ranges
  • Perform statistics for efficient trend and production analysis
  • Export data to PC in various formats including (XML, CSV, XLSX, or PDF)
  • Ideal for MS-TS and ML-T balances
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Advanced and Standard Level Balances
Connection Type Ethernet or RS232
For Use With (Application) Collecting Weighing Data
Includes 3 Licenses
Type Software License