Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ 1,1,1,3,3,3-Hexafluoro-2-Propanol, MilliporeSigma™ Supelco™

Catalog No. 111022433 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2433 50 mL
11-102-2432 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-2433 Supplier MilliporeSigma Supelco Supplier No. 1812750ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 920-66-1
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.04
MDL Number MFCD00011651
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym HFP; Hexafluoroisopropanol
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES OC(C(F)(F)F)C(F)(F)F

Specifications

Melting Point -4°C (literature)
Density 1.596 g/mL (at 25°C (literature))
Boiling Point 59°C (literature)
Flash Point Closed Cup (Boils Before Flash)
Refractive Index n20/D 1.275 (literature)
Linear Formula (CF3)2 CHOH
Quantity 50 mL
Formula Weight 168.04
Percent Purity ≥99.8% (GC)
Hazard Category H314 - H361fd - H373
RTECSNumber UB6450000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.