Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ 2,3,4,5,6-Pentafluorobenzyl bromide, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019340 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9340 5 g
11-102-0438 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9340 Supplier MilliporeSigma Supelco Supplier No. 902575G
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00000299
Synonym PFB-Br; alpha-Bromo-2,3,4,5,6-pentafluorotoluene; (Bromomethyl)pentafluorobenzene

Specifications

Melting Point 19°C to 20°C
CAS 1765-40-8
Density 1.728 g/mL (at 25°C)
Boiling Point 174°C to 175°C
Linear Formula C6F5 CH2 Br
Quantity 5 g
Formula Weight 260.99
Percent Purity ≥98.5% (GC)
Grade For GC derivatization
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.