Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Boron trichloride Solution, 12% in Methanol, For GC Derivatization, MilliporeSigma™ Supelco™

Catalog No. 111019225 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
10 x 1 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9225 100 mL
11-101-9271 10 x 1 mL
11-101-8416 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-9225 Supplier MilliporeSigma Supelco Supplier No. 91379100ML
Add to Cart
Add to Cart

Specifications

CAS 10294-34-5
Quantity 100 mL
Linear Formula BCl3
MDL Number MFCD00011313
Formula Weight 117.17
Grade GC Derivatization
Product Line LiChropur
Density 0.875-0.895 g/mL (at 20°C)

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.