Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Dibutylammonium acetate Solution, 0.5 M in Water, MilliporeSigma™ Supelco™

Catalog No. 111013408 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
6 x 25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3408 6 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3408 Supplier MilliporeSigma Supelco Supplier No. 733456EA
Add to Cart
Add to Cart

Dibutylammonium acetate is an ion-pairing agent with quaternary ammonium ions. It generally helps in ionization of carbohydrate fragments providing a volatile solvent for the mass spectrometric technique.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00191381
Synonym Dibutylamine acetate Concentrate

Specifications

CAS 19070-91-8
Packaging Ampule
Linear Formula (CH3 CH2 CH2 CH2)2 NH · CH3 COOH
Quantity 6 x 25 mL
Formula Weight 189.30
Grade For ion pair chromatography
Concentration or Composition (by Analyte or Components) 0.5 M aq. soln.
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.