Learn More
LiChropur™ Hexadecyltrimethylammonium bromide, ≥99.0% (AT), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Melting Point | 248°C to 251°C |
| Physical Form | Powder or Crystals |
| CAS | 57-09-0 |
| Quantity | 50 g |
| Linear Formula | CH3(CH2)15 N(Br)(CH3)3 |
| Flash Point | 244°C |
| Solubility Information | Water: soluble 36.4 g/L (at 20°C (completely)) |
| Formula Weight | 364.45 |
| Percent Purity | ≥99.0% (AT) |
| Grade | Ion Pair Chromatography |
| Show More |
Safety and Handling
| RTECSNumber | BQ7875000 |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.