Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Methoxyamine Hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111022797 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2797 1 g
11-102-2798 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-2797 Supplier MilliporeSigma Supelco Supplier No. 898031G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methoxyamine hydrochloride is a commonly used reagent for the generation of oxime derivatives of aldehyde- or ketone-group containing compounds. It is typically employed as an oximation reagent prior to silylation derivatizations.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.52
MDL Number MFCD00012951
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym O-Methylhydroxylamine hydrochloride; Methoxylamine hydrochloride
IUPAC Name hydrogen O-methylhydroxylamine chloride
SMILES [H+].[Cl-].CON

Specifications

Melting Point 151°C to 154°C (literature)
Linear Formula CH3ONH2 · HCl
Quantity 1 g
UN Number UN3261 - Class 8 - PG 2 - EHS - Corrosive solid, acidic, organic, n.o.s., HI: all
Formula Weight 83.52
Percent Purity 97.5% to 102.5% (AT)
Grade For GC Derivatization
Hazard Category H290 - H302 + H312 - H315 - H317 - H319 - H351 - H372 - H410

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.