Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ N-Methyl-N-trimethylsilylheptafluorobutyramide, 90%, MilliporeSigma™ Supelco™

Catalog No. 111018336 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8336 5 mL
11-101-9233 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8336 Supplier MilliporeSigma Supelco Supplier No. 694845ML
Add to Cart
Add to Cart

Specifications

CAS 53296-64-3
Product Line LiChropur
Density 1.254 g/mL (at 25°C)
Flash Point 33°C - closed cup
Linear Formula CF3 CF2 CF2 CON(CH3)Si(CH3)3
MDL Number MFCD00077822
Boiling Point 148°C
Synonym N-Trimethylsilyl-N-methylheptafluorobutyramide; MSHFBA
Type Organic Standard
Quantity 5 mL
Formula Weight 299.26
Percent Purity ≥90% (GC)
Grade GC Derivatization
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.