Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, ≥97.5-98%, MilliporeSigma™ Supelco™

Catalog No. 111018220 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8220 25 mL
11-102-0404 5 mL
11-101-8465 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8220 Supplier MilliporeSigma Supelco Supplier No. 1520925ML
Add to Cart
Add to Cart

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane has found application to be used in gas chromatography (GC) and mass-spectrometry (MS). It can also be used in HPLC- electrochemical detection assay of 8-oxo-deoxyguanosine and?8-oxo-guanine.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00008269
Synonym BSTFA + TMCS

Specifications

CAS 25561-30-2
Density 0.97 g/mL
Boiling Point 45°C to 50°C (14 mmHg)
Flash Point -9°C
Packaging Ampule
Linear Formula CF3 C[=NSi(CH3)3 ]OSi(CH3)3
Quantity 25 mL
Formula Weight 257.40
Percent Purity ≥97.5% (excluding TMCS, GC); 98% (excluding TMCS)
Grade For GC derivatization
Product Line LiChropur
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.