Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, ≥98.5-99%, MilliporeSigma™ Supelco™

Catalog No. 111018072 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8072 100 mL
11-102-0670 1 mL
11-102-0237 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8072 Supplier MilliporeSigma Supelco Supplier No. 15238100ML
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00008269
Synonym Silylating mixture V; BSTFA + TMCS

Specifications

CAS 25561-30-2
Density 0.97 g/mL
Boiling Point 45°C to 50°C (14 mmHg)
Flash Point 6°C
Packaging Glass Bottle
Linear Formula CF3 C[=NSi(CH3)3 ]OSi(CH3)3
Quantity 100 mL
Formula Weight 257.40
Percent Purity ≥98.5% (excluding chlorotrimethylsilane, GC); 99% (excluding TMCS)
Grade For GC derivatization
Product Line LiChropur
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.