Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, 99% (excluding TMCS), MilliporeSigma™ Supelco™

Catalog No. 111018800 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mL
10 x 0.1 mL
10 x 1 mL
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-101-8800 10 x 1 mL Glass Bottle
11-101-9003 10 x 0.1 mL Glass Bottle
11-101-8739 25 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8800 Supplier MilliporeSigma Supelco Supplier No. 1523810X1ML
Only null left
Add to Cart
Add to Cart

For GC Derivatization, Contains 1% TMCS

Silylating mixture used for the silylation of various compounds
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.40
MDL Number MFCD00008269
InChI Key XCOBLONWWXQEBS-UHFFFAOYSA-N
Synonym Silylating mixture V; BSTFA + TMCS
IUPAC Name trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
SMILES C[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F

Specifications

Density 0.97 g/mL
Boiling Point 45°C to 50°C (14 mmHg)
Packaging Glass Bottle
Linear Formula CF3C[=NSi(CH3)3 ]OSi(CH3)3
Refractive Index n20/D 1.384
Quantity 10 x 1 mL
Formula Weight 257.40
Percent Purity 99% (excluding TMCS)
Grade For GC derivatization
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.