Learn More
LiChropur™ Pentafluoropropionic anhydride, 99%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 356-42-3 |
| Density | 1.571 g/mL (at 25°C) |
| Boiling Point | 69°C to 70°C (735 mmHg) |
| Packaging | Ampule |
| Linear Formula | (CF3 CF2 CO)2 O |
| Quantity | 100 mL |
| Formula Weight | 310.05 |
| Percent Purity | 99% |
| Grade | For GC derivatization |
| Product Line | LiChropur |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.