Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Pentafluoropropionic anhydride, 99%, MilliporeSigma™ Supelco™

Catalog No. 111018398 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8398 25 mL
11-101-9174 5 mL
11-101-8609 10 x 1 mL
11-101-7965 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-101-8398 Supplier MilliporeSigma Supelco Supplier No. 39490425ML
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00000429
Synonym PFPA; Perfluoropropionic anhydride

Specifications

CAS 356-42-3
Density 1.571 g/mL (at 25°C)
Boiling Point 69°C to 70°C (735 mmHg)
Packaging Ampule
Linear Formula (CF3 CF2 CO)2 O
Quantity 25 mL
Formula Weight 310.05
Percent Purity 99%
Grade For GC derivatization
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.