Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Potassium phosphate dibasic, For HPLC, 99.0-101.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111015511 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5511 250 g
11-101-5512 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5511 Supplier MilliporeSigma Supelco Supplier No. 59678250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
MDL Number MFCD00011383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym Dipotassium hydrogenphosphate; Dipotassium phosphate; sec.-Potassium phosphate
IUPAC Name dipotassium hydrogen phosphate
SMILES [K+].[K+].OP([O-])([O-])=O

Specifications

Quantity 250 g
Linear Formula K2HPO4
Formula Weight 174.18
Percent Purity 99.0 to 101.0% (T)
Grade HPLC
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.