Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Sodium Perchlorate Monohydrate, MilliporeSigma™ Supelco™

Catalog No. 111022671 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2671 250 g
11-102-2672 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-2671 Supplier MilliporeSigma Supelco Supplier No. 56581250G
Add to Cart
Add to Cart

For HPLC

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7-3-7791
Molecular Formula ClH2NaO5
Molecular Weight (g/mol) 140.45
MDL Number MFCD00149164
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
IUPAC Name sodium hydrate perchlorate
SMILES O.[Na+].[O-][Cl](=O)(=O)=O

Specifications

pH 6 to 8
Quantity 250 g
Linear Formula NaClO4 · H2O
Solubility Information H2O: 5%, Clear, Colorless
Formula Weight 140.46
Percent Purity 99% to 101% (T)
Grade HPLC
Hazard Category H271
RTECSNumber SC9850000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.