Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Triethylamine, For HPLC, 99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111017017 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Packaging:
Clear Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-101-7017 2 mL Clear Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7017 Supplier MilliporeSigma Supelco Supplier No. 811012ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for use as a mobile phase in HPLC applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00009051
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym N,N-Diethylethanamine
IUPAC Name triethylamine
SMILES CCN(CC)CC

Specifications

Melting Point -115°C
Density 0.726 g/mL (at 25°C)
Boiling Point 88.8°C
Packaging Clear Glass Bottle
Linear Formula (C2H5)3 N
Refractive Index n20/D 1.401
Quantity 2 mL
Vapor Density 3.5 (vs. air)
Formula Weight 101.19
Vapor Pressure 51.75 mmHg (20°C)
Percent Purity ≥99.5% (GC)
Grade For HPLC
Physical Form Liquid
Product Line LiChropur
Show More Show Less
RTECSNumber YE0175000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.