CAS RN 121-44-8
Triethylamine (HPLC), Fisher Chemical™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.19 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N IUPAC Name: triethylamine SMILES: CCN(CC)CC
Triethylamine, 99%
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 MDL Number: MFCD00009051 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine 99.0+%, TCI America™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 MDL Number: MFCD00009051 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, for HPLC
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.19 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, 99%, pure
CAS: 121-44-8 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, HPLC, >99.5%, MilliporeSigma™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.19 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,triethyl amine,trietilamina,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026
Triethylamine (Reagent), Fisher Chemical™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 MDL Number: MFCD00009051 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, 99.5%, for analysis
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.19 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, 99.7%, extra pure
CAS: 121-44-8 MDL Number: MFCD00009051 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, MilliporeSigma™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, BAKER BIO-ANALYZED™ Reagent, J.T. Baker™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, MilliporeSigma™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
LiChropur™ Triethylamine, For HPLC, 99.5% (GC), MilliporeSigma™ Supelco™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.19 MDL Number: MFCD00009051 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: N,N-Diethylethanamine IUPAC Name: triethylamine SMILES: CCN(CC)CC
Triethylamine OR, Macron Fine Chemicals™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
- 1
- 2