Learn More
Supelco Inc RP-18 HPLC Column
Supplier:  Supelco Inc 50137U
Encompass
LiChrospher RP-18 HPLC column (L × I.D. 25 cm × 4 mm) is found to be reliable and high versatile. Suitable for reversed-phase high performance liquid chromatography (RP-HPLC) of acidic, neutral and weakly basic compounds, according to L1 USP specifications.
Catalog No. NC1844149
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.