Learn More
Apexbio Technology LLC Lidocaine, 5g Cas# 137-58-6
SDP
Supplier:  Apexbio Technology LLC A14505000
Encompass_Preferred
Anasthetic and class Ib antiarrhythmic agent.?Blocks voltage-gated sodium channels in the inactivated state. Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8510
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.