Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lidocaine Hydrochloride, Monohydrate, USP, 97.5-102.5%, Spectrum™ Chemical
SDP

Catalog No. 18603871 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5 kg
10 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-871 25 kg Fiber Drum
18-603-867 25 g Amber Glass Bottle
18-603-868 100 g Amber Glass Bottle
18-603-869 500 g Amber Glass Bottle
18-603-873 5 kg Poly Pail
18-603-870 10 kg Poly Pail
18-603-872 50 kg Fiber Drum
18-603-866 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-603-871 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. LI10325KGBL
Add to Cart
Add to Cart
Request a Quote

LI103, 6108-05-0, C14H22N2O·HCl·H2O

Spectrum™ Chemical Lidocaine Hydrochloride, Monohydrate, USP is used as a topical local anesthetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6108-05-0
Molecular Formula C14H25ClN2O2
Molecular Weight (g/mol) 288.82
MDL Number MFCD00150329
InChI Key YECIFGHRMFEPJK-UHFFFAOYSA-N
IUPAC Name {[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride
SMILES O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C

Specifications

CAS Max % 100%
Packaging Fiber Drum
Quantity 25 kg
Percent Purity 97.5 to 102.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.