Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco Life/form™ Advanced Sanitary CPR Dog
SDPSCI_ED

Catalog No. S00109 Shop All Simulaids, Inc. Products
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Animal Medical Practice
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S00109 Animal Medical Practice
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00109 Supplier Nasco Supplier No. LF01155
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Life/form™ Advanced Sanitary CPR Dog provides each student with their own sanitary muzzle and nosepiece with a one-way valve.

  • The Life/form™ Advanced Sanitary CPR Dog provides each student with their own sanitary muzzle and nosepiece with a one-way valve.
  • The muzzles may be disinfected and reused, while the nosepieces and lower airways are disposable.
  • In addition to practice in assisted breathing, students may practice the appropriate rate, pressure, and position of cardiac massage and coordinate the respiratory cardiovascular functions. An IV can even be established for more advanced training.
  • The Life/form™ Advanced CPR Dog is connected to a small box that displays a light when the correct ventilation volume is applied, another light indicates correct position for cardiac massage, a third light indicates correct compression depth, and an audible beep and light indicates excessive pressure is being applied. A visible chest rise appears when correct ventilations are applied.
  • Includes 10 disposable nosepieces, three disposable lower airways, endotracheal tube, lubricant, 12 cc syringe, towels, butterfly set, artificial blood, two IV bags, control box with AC adapter, 9V battery, and soft carry case.
  • Five-year warranty.

Specifications

Product Type Advanced Sanitary CPR Dog
For Use With (Application) Animal Medical Practice
Color Black
Includes 10 disposable nosepieces, three disposable lower airways, endotracheal tube, lubricant, 12 cc syringe, towels, butterfly set, artificial blood, two IV bags, control box with AC adapter, 9V battery, and soft carry case.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.