Learn More
Nasco Life/form™ Advanced Venipuncture and Injection Arm
SDPSCI_ED
Catalog No. S97903 Shop All Simulaids, Inc. Products
Change view
Click to view available options
Color:
Black skin
White Skin
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S97903 White Skin
S97904 Black skin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S97903 Supplier Nasco Supplier No. LF01121
Add to Cart
Add to Cart
Includes
Two quarts simulated blood, 3 cc syringe, 12 cc syringe, needle, two fluid bags, instruction manual and a carrying case
A revolutionary training arm providing complete venous access for IV therapy and phlebotomy, as well as sites for intramuscular and intradermal injections.
An extensive 8-line vascular system allows students to practice venipuncture at all primary and secondary locations, including starting IVs and introducing IV catheters.
  • One external fluid bag supplying artificial blood to all veins simultaneously
  • Dorsal surface includes injectable metacarpal, digital and thumb veins
  • Antecubital fossa includes the median cephalic, median basilic and median cubital veins
  • Can be performed along with basilic, cephalic, accessory cephalic and median antebrachial veins
  • Intramuscular injections may be performed in the deltoid muscle
  • Intradermal injection sites are located in the upper arm
  • Intramuscular injections into the deltoid muscle are enhanced by the soft, lifelike skin and by the natural bony landmarks in the region
  • Complete replacement kits are available and easy to use
  • Soft, flexible fingers are molded separately with extreme attention
  • Flexion of the wrist helps students develop manipulation skills
  • Replaceable skin rolls as the veins are palpated and a discernable pop is felt when entering the veins
  • Life/form molding process reproduces the fine details of the skin to make the arm look and feel alive
  • Valves in the veins can be seen and palpated on the skin surface
  • To extend the life of the veins, an aerosol sealant is available to seal punctures and prevent leakage
Warranty and Services
Three years
Specifications
Product Type Advanced Venipuncture and Injection Arm
For Use With (Application) Revolutionary Training Arm Providing Complete Venous Access for IV Therapy and Phlebotomy, as well as Sites for Intramuscular and Intradermal Injections
Color White Skin
Includes Two quarts simulated blood, 3cc syringe, 12cc syringe, needle, two fluid bags, instruction manual and a carrying case