Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco Life/form™ Canine IV Leg
SDPSCI_ED

Catalog No. S00110 Shop All Simulaids, Inc. Products
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Canine IV Training
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S00110 Canine IV Training
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00110 Supplier Nasco Supplier No. LF01016
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • The Life/form™ Canine IV Leg represents the left foreleg of a medium-sized dog in the sternal recumbent position, and features two interchangeable sleeves - normal fur for routine blood draws, and a shaved area for IV catheter insertion.
  • The replaceable vein system demonstrates actual blood flashback. Great for bandaging and restraint/assistance practice as well.
  • Includes a portable storage box with built-in IV pole, blood powder, syringe, IV catheter, and two fluid supply bags

Specifications

Product Type Medical Model
For Use With (Application) Canine IV Training
Color Black
Includes Portable storage box with built-in IV pole, blood powder, syringe, IV catheter, and two fluid supply bags.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.