Learn More
LIFE™ O2 SoftPac™ Portable Emergency Oxygen Units
Catalog No. 19097464 Shop All Life Corporation Products
Click to view available options
Product Type:
Emergency Oxygen Unit
Fixed Flow Softpac Emergency Oxygen Unit
Variable Flow Softpac Emergency Oxygen Unit
Volume (Metric) Reservoir:
246 L
250 L
255 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Volume (Metric) Reservoir
19-097-464 Fixed Flow Softpac Emergency Oxygen Unit 250 L
19-097-465 Variable Flow Softpac Emergency Oxygen Unit 246 L
19-097-466 Emergency Oxygen Unit 255 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-097-464 Supplier LIFE™ Supplier No. LIFE2612
Add to Cart
Add to Cart
Life™ O2 SoftPac™ Portable Emergency Oxygen Units are a light-weight, easily portable unit for frequent carrying by first responders to provide emergency oxygen to a breathing victim or oxygen-enriched CPR to a non-breathing victim.
  • 6 pounds
  • Constant reading supply gauge visible through a clear window
  • Shoulder strap, foam handle, and padded case for easy carrying
  • Wall mount hook
  • Universally fits both adults and children with one-way valve and filter to prevent disease transmission
  • Disposable/replaceable (or refillable) cylinder
  • Non-prescription
  • 40-minute supply at 6 LPM
  • Shipped with cylinder full of O2 and ready to use
Specifications
Product Type Fixed Flow Softpac Emergency Oxygen Unit
Material Aluminum
For Use With (Application) Serves as inhalator for breathing victim, resuscitator for nonbreathing victim
Includes Handle and shoulder strap, wall mount hook and bag
Quantity 1 Ea.
Volume (Metric) Reservoir 250 L
Weight (English) 6 lb.
Weight (Metric) 2.72 kg