Learn More
Walter Products™ Life-size Dual-sex Human Torso with Open Back
SCI_ED
Catalog No. S139137 Shop All Walter Products Products
Change view
Click to view available options
Dimensions (L x W x H):
13 x 10 x 34 in.
Catalog No. Dimensions (L x W x H)
S139137 13 x 10 x 34 in.
1 options
This item is not returnable. View return policy
Catalog No. S139137 Supplier Walter Products™ Supplier No. B10108
Add to Cart
Add to Cart
This item is not returnable. View return policy
This life-size human torso model contains 27 removable parts, providing students with a hands-on experience examining main body systems.
  • Removable parts feature exchangeable male and female reproductive organs
  • Head is opened, exposing one half of the brain
  • Neck is dissected to show muscular, glandular, vascular, and neural structures
  • Cerebellum and lungs can be completely removed, showing the important structures of the head and inner wall of the thoracic cavity
  • Thorax and abdomen are removable, allowing easy observation of the internal organs
  • Back is opened and dissected showing the vertebral column, spinal cord, and nerve endings
  • Further examination of the spinal cord is also possible with the removal of the 12th thoracic vertebrae
  • 27 Parts Include: Torso, Cerebrum and Cerebellum, Eyeball, Lung (4 parts), Heart (2 parts), Liver, Esophagus, Thoracic Aorta, Half of Kidney, Pancreas, Spleen, Duodenum, Stomach (2 parts), Large and Small Intestine, Cecum Cover, Male and Female Organs (8 Parts), 12th Thoracic Vertebra with Spinal Cord
Specifications
Dimensions (L x W x H) 13 x 10 x 34 in.
Material PVC
Product Type Anatomical Model