Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Life Size Flexible Skeleton Model with Muscles
SCI_ED

Catalog No. S81002 Shop All Walter Products Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S81002 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81002 Supplier Walter Products™ Supplier No. B10230
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This full-size human skeleton has a flexible spine that can bend to mimic natural human movement

  • Life-size human skeleton made of high quality medical PVC
  • Skeleton has flexible spine to mimic natural movement
  • Accurately painted and numbered, showing the muscle origins in red and muscle insertions in blue
  • Skull has movable jaw, removable skull cap, suture lines and a full dentition
  • Both arms and legs can be removed for closer study
  • Pelvic mounted on a roller base and supplied with a diagram

Specifications

For Use With (Application) Education
Height (Metric) 170 cm
Includes Skeleton & roller stand
Material PVC
Quantity 1
Size Life-Size
Weight (Metric) 11.79 kg
Product Type Model, Skeleton, Flexible with Muscles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.