Learn More
Walter Products™ Life-size Flexible Spinal Column Model
SCI_ED
Catalog No. S139147 Shop All Walter Products Products
Change view
Click to view available options
Material:
PVC
Catalog No. Material
S139147 PVC
1 options
Catalog No. S139147 Supplier Walter Products™ Supplier No. B10245
Add to Cart
Add to Cart
Life-size and fully flexible for a greater understanding of spinal movement, this model includes color-coded regions and muscles.
  • Life-size vertebral column includes the sacrum, coccyx, pelvis, and removable femur heads
  • Complete with the pubic symphysis, this model shows great detail representing the vertebral arteries, spinal nerve branches, and intervertebral discs
  • Muscles origins (red) and insertions (blue) are also depicted on the left side of the model for further learning
  • Model shows the 5 different sections of the spinal column: 7 cervical vertebrae, 12 thoracic vertebrae, 5 lumbar vertebrae, including prolapsed L3-L4 discs sacrum coccyx
  • Comes with a hanging stand and labeled diagram
Specifications
Material PVC
Product Type Anatomical Model