Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Life-Size Human Skeleton Model with Dust Cover
SCI_ED

Catalog No. S80999 Shop All Walter Products Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S80999 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80999 Supplier Walter Products™ Supplier No. B10201HD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Life size skeleton with exclusive dust cover.

  • First class natural cast
  • Life size 170 cm, high quality
  • Skull is 3-part. Arms and legs are flexible for human movements
  • Color chart included
  • Heavy duty dust cover included

Specifications

For Use With (Application) Education
Height (Metric) 170 cm
Includes Skeleton, roller base, hanging rod, diagram, and dust cover
Material PVC
Quantity 1
Size Life-size
Weight (Metric) 12
Product Type Model, Human Skeleton
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.