Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Life-Size Human Stomach Model
SCI_ED

Catalog No. S24454 Shop All Eisco Products
Change view
Click to view available options
L x W x H: 7 x 3 x 6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S24454 L x W x H: 7 x 3 x 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24454 Supplier Eisco™ Supplier No. AM288AS

missing translation for 'validMainframeMsg'

Highly detailed demonstration model of internal structures of stomach.

  • Bisectable internal and external structures
  • Can be removed from rod base
  • Includes key card with 25 call-outs
  • Base mounted

Specifications

Dimensions (L x W x H) 7 x 3 x 6 in.
For Use With (Application) Demonstration of internal structures of stomach
Includes key card with 25 call-outs
Material Plastic
Mounting 5 x 5 in. Rod Base
Size Life-Size
Product Type Model, Stomach, Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.