Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Calibration Kit, Fast 96-well

Catalog No. 4432563
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 96-well plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-325-63 1 x 96-well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-325-63 Supplier Applied Biosystems™ Supplier No. 4432563
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Region of interest (ROI) plate, Background plate, Dye calibration plates, and RNase P instrument verification plate

Used to maintain your real-time PCR instrument with Fast 96-well block.

The kit includes region of interest (ROI), background, and dye calibrations, as well as RNase P verification. Please see your instrument user guide for full instructions on use.

See instrument user guide for full instructions.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Region-of-Insert (ROI)
Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Product Type Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 x 96-well plate
Calibration Type Spectral Calibration

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.