Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cassette Clamp Cam Handle Set

Catalog No. A26732
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26732 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26732 Supplier Invitrogen™ Supplier No. A26732

Retrofit part for the Bolt™ Mini Gel Tank

The Cassette Clamp Cam Handle Set is a retrofit part for the Bolt™ Mini Gel Tank to enable it to securely fit Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels in a manner similar to the Mini Gel Tank. Customers who have the Bolt™ Mini Gel Tank can replace the original black cam handles on their cassette clamps with these grey cam handles to more conveniently run these 10 cm wide gels.

About the Mini Gel Tank
The Mini Gel Tank is compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
For Use With (Equipment) Bolt™ Mini Gel Tank
Gel Compatibility Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels
Gel Size Mini
Type Cassette Clamp Cam Handle Set
Gel Dimensions 10 cm wide
Quantity 1 unit
Shipping Condition Room Temperature
Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

How can I upgrade the Bolt Mini Gel Tank to run the Bolt Bis-Tris Plus gels I recently purchased?

The Bolt Mini Gel Tank has been discontinued as of December 31, 2014, and will be offered until inventory is depleted. To upgrade the Bolt Mini Gel Tank to run the new Bolt Bis-Tris Plus gels as well as other Invitrogen Mini gels, you have to replace the black 10.5 cm cassette clamp cam handles with gray 10 cm cassette clamp cam handles (Cat. No. A26732). Instructions for replacement of the cam handles can be found in this video.

Can I purchase replacement parts for the Mini Gel Tank?

Here are the replacement parts we offer for the Mini Gel Tank:
Replacement part - Cat. No.
Gel Runner Tank - B4478641
Mini Gel Tank Lid - A25944
Mini Gel Tank Base - A25950
Cassette Clamp, left - A25946
Cassette Clamp, right - A25945
Cassette Clamp Cam Handle Set - A26732

What are the components of the Mini Gel Tank?

The Mini Gel Tank includes the Mini Gel Electrophoresis Tank, Mini Gel Tank Lid, Cassette Clamp (Right), 10 cm, Cassette Clamp (Left), 10 cm, Mini Gel Tank Base, Invitrogen Power Supply Adapters, and Gel Knife.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.