Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ ViiA™ 7 Array Card Spectral Calibration Kit

Catalog No. 4432314
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-323-14 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-323-14 Supplier Applied Biosystems™ Supplier No. 4432314
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

Includes

Empty array cards and seven tubes of calibration dyes including: FAM™ dye, VIC™ dye, ROX™ dye, ROI dye mix, background buffer, FAM/ROX dye, VIC/ROX dye

Perform spectral calibration, region-of-interest (ROI) calibration and normalization on the ViiA™ 7 or QuantStudio™ 7 Flex or 12K Flex systems with array card block. Applied Biosystems™ ViiA™ 7 Array Card Spectral Calibration Kit includes empty array cards and seven tubes of calibration dyes.

It includes empty array cards and seven tubes of calibration dyes including: FAM™ dye, VIC™ dye, ROX™ dye, ROI dye mix, background buffer, FAM™/ROX™ dye, VIC™/ROX™ dye. Please see your instrument maintenance guide for full instructions on use. The dye formulations for your ViiA™ 7, QuantStudio™ 7, or QuantStudio™ 12K Flex system will improve results with multiplexing by more accurately representing fluorescent spectra found in your real-time PCR experiments.

The Applied Biosystems™ ViiA™ 7 Array Card Spectral Calibration Kit includes empty array cards and seven tubes of calibration dyes including: FAM™ dye, VIC™ dye, ROX™ dye, ROI dye mix, background buffer, FAM™⁄ROX™ dye, VIC™⁄ROX™ dye.

The dye formulations for the ViiA™ 7, QuantStudio™ 7, or QuantStudio™ 12K Flex system will improve results with multiplexing by more accurately representing fluorescent spectra found in real-time PCR experiments.

Order Info

Store plates at -15° to -25°C

TRUSTED_SUSTAINABILITY

Specifications

Format 384-well Microfluidic Card
Content And Storage Store 7 Tubes of Calibration Dyes at -15°C to -25°C
Calibrated Dye FAM, ROX, VIC
For Use With (Equipment) QuantStudio™ 12 K Flex, QuantStudio™ 7, ViiA™ 7 System
Product Line ViiA
Product Type Array Card Spectral Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 kit
Calibration Type Spectral Calibration, Region-of-Interest (ROI) Calibration, Normalization
I cannot open the QuantStudio 3/5 Design & Analysis 2 (DA2) software on my Mac. What are the software requirements for Mac?

To open the software download on your Mac, you will need to adjust your security settings using the following instructions from Apple support:

Safely open apps on your mac

Please note, the software should run on Mac OS 10.10 or later.

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

Can I import a standard curve for my real-time PCR data into the QuantStudio Real-Time PCR Software?

If you are analyzing results from the ViiA 7 Real-Time PCR System, QuantStudio 6 Flex Real-Time PCR System, or QuantStudio 7 Flex Real-Time PCR System, it is possible to import a standard curve into the QuantStudio Real-Time PCR Software following the instructions below:
1. Open the data file in the QuantStudio Real-Time PCR Software.
2. Go to Analysis Settings.
3. Click on the "Standard Curve Settings" tab.
4. Select the option to "Use a standard curve imported from another experiment" and click on "Import".
5. Browse to and highlight the proper data file and click "Select".
6. Click Apply Analysis Settings.

How can I get a summary of the calibrations performed on the QuantStudio 6 and 7 Flex Real-Time PCR Systems?

You can get a summary of the calibrations from the QuantStudio Software by going to Instrument > Instrument Console > Manage Instrument > Information > Print Calibration Log.

Do I need to back up my calibration files before I upgrade the software for the ViiA7 Real-Time PCR System?

No, you do not need to back up the calibration files as all of the calibration files are stored on the ViiA7 Real-Time PCR instrument. Once the computer software is properly installed and the computer is connected to the instrument, the calibration information will automatically update in the QuantStudio Software.

Can I still start a run on the ViiA7 Real-Time PCR System if the "Calibration is expired" notification is displayed?

Yes, you can still start a run on the ViiA7 Real-Time PCR System if the expired calibration notice displays. This is just a reminder and you can close it to proceed with the run. Additionally, you can change when you want the notifications to show up and how frequently you want to be notified to calibrate the instrument. To do this, go to the toolbar in the QuantStudio Software and select Instrument > Instrument Console. Select your instrument icon and click on Manager Instrument > Maintenance > Calibration Reminders. From the Calibration reminders Settings page you can update when the calibrations expire, turn expiration reminders on/off, and set when the reminders show up.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.