Learn More
Selleck Chemical LLC Lifirafenib (BGB-283) 5mg 1446090-79-4 Beigene-283
SDP
Supplier:  Selleck Chemical LLC S79265MG
Encompass_Preferred
Lifirafenib (BGB-283, Beigene-283) potently inhibits RAF family kinases and EGFR activities in biochemical assays with IC50 values of 23, 29 and 495 nM for the recombinant BRAFV600E kinase domain, EGFR and EGFR T790M/L858R mutant. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8120
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.