Learn More
CHROMAGENIX LLC LT Human IgA Kit w/ Microspin columns
SDP
Supplier:  CHROMAGENIX LLC LT150KIT
Encompass_Preferred
LigaTrap Human IgA Purification Kit includes ten 0.100mL microspin columns and buffers for small volume (microliters) and follow a "batch" purification method. This product is based on proprietary LigaTrap technology to utilize novel, patented ligands, designed for optimized antibody purification.
Catalog No. 50-201-6107
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.