Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Light Leaper Demonstrator
SDPSCI_ED

Catalog No. S27825 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S27825 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27825 Supplier United Scientific™ Supplier No. LLA001
Add to Cart
Add to Cart

Use the Light Leaper Demonstrator to connect to an electrostatic generator to create simultaneous sparks in each break as charge flows.

  • Popular accessory for electrostatic demonstrations.
  • Connect to an electrostatic generator to create simultaneous sparks in each break as charge flows.
  • Only compatible with electric Van de Graaff Generators.

Specifications

Grade 9 to 15
Age 14 to 20 Yr.
For Use With (Equipment) Electric Van de Graaff Generators
Includes An insulating printed circuit board with six breaks and a binding post at each end.
Class Size 1 Lab Group
Product Type Light Leaper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.