Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Light Mineral Oil, NF, Spectrum™ Chemical
SDP

Catalog No. 18603893 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-893 20 L Amber Glass Bottle
18-603-889 500 mL Amber Glass Bottle
18-603-890 1 L Amber Glass Bottle
18-603-891 4 L Amber Glass Bottle
18-603-892 4 x 4 L Amber Glass Bottle
18-603-894 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-603-893 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M129320LT
Add to Cart
Add to Cart

M1293, 8042-47-5

Spectrum™ Chemical Light Mineral Oil, NF is used as a vehicle and humectant in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8042-47-5

Specifications

CAS Max % 100%
Boiling Point >230°C / 446°F
Packaging Amber Glass Bottle
Quantity 20 L
Grade BP, EP, FCC, JP, USP
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Light Mineral Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.