Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ 2D Light Module for Gbox XX6 and XX9
GREENER_CHOICESDP

Catalog No. 01257184 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Gbox Chemi XX6/9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-257-184 Gbox Chemi XX6/9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-257-184 Supplier Syngene™ Supplier No. DYVLIGHTMOD
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Syngene™ 2D Light Module for Gbox XX6 and XX9 is a CY dye edge light module(28x21cm) with low fluorescent plates for capture of 2D DIGE gels.

  • Required for multiplexing CY dyes
  • Contains Red, Green and Blue multiplex lights

Specifications

For Use With (Application) 2d DIGE electrophoresis imaging
For Use With (Equipment) Gbox Chemi XX6/9
Light Source LED
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.