Learn More
Active Motif LightSwitch™ Luciferase Assay Kit
SDP
Supplier:  Active Motif 32031
Encompass_Preferred
Active Motif's LightSwitch Luciferase Assay System is comprehensive system for studying transcriptional and post-transcriptional regulation. It includes over 30,000 pre-cloned promoter and 3'UTR sequences that have been cloned from the human genome, which are assayed using optimized reagents and protocols that make every step of the procedure faster and simpler. The LightSwitch Luciferase Assay Kit contains a novel, proprietary assay substrate that was formulated to provide high sensitivity and a broad dynamic range when assaying the engineered RenSP luciferase gene found in all LightSwitch reporter constructs.
Catalog No. NC0999256
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.