Learn More
Cole-Parmer AEMC CA813 High-range light meter with mountable pensor
Supplier:  Cole-Parmer 8653310
Encompass_Preferred
Light meters feature optical sensors designed to match the response of the human eye—ideal for assessment of employee workspace light levels. Powerful meters feature an ergonomic housing, large display with HOLD option, and intuitive setting controls. Both models include a shockproof protective holster to protect meter from dirt and damage.
Catalog No. 50-992-652
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.