Description
- Simple conjugation workflow with minimal hands-on time.
- Labeled antibodies can be used in western blotting and ELISA without further purification.
Specifications
Specifications
| Quantity | 100 μg |
| Product Type | Antibody Labeling Kit |
| Content And Storage | -20°C |
| Label Type | Coumarin Dyes |
| Product Line | Novus |