Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) HRP Antibody Labeling Kit
SDP

Catalog No. 7010000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
3 x 10 μg Reaction
3 x 100 μg Reaction
5 mg
5 x 1 mg Reaction
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-100-00 3 x 100 μg Reaction
70-100-02 1 mg
70-100-03 5 x 1 mg Reaction
70-100-04 5 mg
70-100-30 3 x 10 μg Reaction
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 70-100-00 Supplier Novus Biologicals™ Supplier No. 7010000
Add to Cart
Add to Cart

Applications: Western Blot, Immunohistochemistry, In vivo assay

Horseradish peroxidase is a 44kDa glycoprotein with 6 lysine residues which can be conjugated to labeled biomolecules using Lightning-Link. The enzyme label can be visualized by chromogenic reactions; for example diaminobenzidine (DAB) in the presence of hydrogen peroxide (H202) is converted in to a water insoluble brown pigment. Other substrates which can be used to measure horseradish peroxidase activity include ABTS, TMB and TMBUS.

Order Info

Quantity

Specifications

Quantity 3 x 100 μg Reaction
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Conjugate HRP
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
For Use With (Application) In vivo assay
Product Type HRP Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.