Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) PE-Cy5.5 Antibody Labeling Kit
SDP

Catalog No. 7610005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
180 μg
30 μg
60 μg
600 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-100-05 60 μg
76-100-10 180 μg
76-100-15 600 μg
76-100-30 30 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 76-100-05 Supplier Novus Biologicals™ Supplier No. 7610005
Add to Cart
Add to Cart

Antibody labeling kit

PE/Cy5.5 is a tandem conjugate. In our Lightning-Link PE/Cy5.5 the best result is obtained when the tandem is excited at 488nm, wavelength at which the PE has its maximum absorption. The excited PE functions as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy5.5. The Cy5.5 emits then the energy received from the PE in the form of long wavelength light at 694nm.

The antibody is covalently bonded to the label in a directional and controlled process at near-neutral pH.In view of the large size of the PE-Cy5.5 label, the amount of antibody used in a labeling reaction with the tandem must always be less than the pack size of LL PE-Cy5.5, in order that the PE-Cy5.5 is in a slight molar excess. The best ratio for any new antibody reagent must be determined by experimentation but 50-60ug of IgG antibody for every 100ug of LL PE-Cy5.5 usually gives optimal results. 50ug of antibody corresponds to an Ab:PE-Cy5.5 molar ratio of 1:1. The amount of antibody used for labeling should be the same as the pack size of LL-PE-Cy5.5. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.

Order Info

Quantity

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 60 μg
Conjugate PE/Cy5.5
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type PE-Cy5.5 Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.