Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) PE-Cy7 Antibody Labeling Kit
SDP

Catalog No. 7620005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
180 μg
60 μg
600 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-200-05 60 μg
76-200-10 180 μg
76-200-15 600 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 76-200-05 Supplier Novus Biologicals™ Supplier No. 7620005
Add to Cart
Add to Cart

Applications: Flow Cytometry

PE/Cy7 is a tandem conjugate. In our Lightning-Link PE/Cy7 the best result is obtained when the tandem is excited at 535nm, wavelength at which the PE has its maximum absorption. The excited PE functions as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy7. The Cy7 emits then the energy received from the PE in the form of long wavelength light at 776nm.

Order Info

Quantity

Specifications

Quantity 60 μg
Conjugate PE/Cy7
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type PE-Cy7 Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.