Learn More
Description
- The Lightning-Link conjugation kit allows PE-Texas Red tandem conjugations to set up in seconds, simply by adding a solution of the antibody to be labeled to a proprietary lyophilised mixture containing PE-Texas Red.
- The amount of antibody used for labeling should be the same as the pack size of LL-PE-Texas Red. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile.
- If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.
PE/TEXAS RED is a tandem conjugate. The PE is excited at 488nm and functions as an energy donor for the TEXAS RED. Energy is transferred from the PE to the TEXAS RED via energy resonance transfer. The TEXAS RED emits the energy received from the PE in the form of long wavelength light at 615nm.
Order Info
Quantity
Specifications
Specifications
| Product Type | PE-Texas Red Antibody Labeling Kit |
| Content And Storage | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent. Store at -20°C. Avoid freeze-thaw cycles. |
| Conjugate | PE/Texas Red |
| Product Line | Novus |
| Quantity | 600 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
