Description
Order Info
Quantity
Specifications
Specifications
| Product Type | PerCP Antibody Labeling Kit |
| Content And Storage | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher reagent. Store at -20°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Quantity | 30 μg |