Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) PerCP Antibody Labeling Kit
SDP

Catalog No. p-7070045 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-800-05 100 μg
71-800-10 300 μg
71-800-30 30 μg
71-800-15 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 71-800-05 Supplier Novus Biologicals™ Supplier No. 7180005
Only null left
Add to Cart
Add to Cart

Applications: Flow Cytometry

Peridinin-chlorophyll-protein complex (PerCP) is a small 35kDa phycobiliprotein isolated from red algae. It has a strong absorption peak at 482nm and a secondary absorption peak at 442nm. PerCP has a large stokes shift (195nm) with its maximum emission peak at 677nm. The protein is commonly used for fluorescent immunolabeling, especially in applications involving fluorescent-activated cell sorting.

Order Info

Quantity

Specifications

Conjugate PerCP
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 100 μg
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher reagent
Product Type PerCP Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.