Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) R-PE Antibody Labeling Kit
SDP

Catalog No. 7030003 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg Reaction
1 x 100 μg Reaction
1 x 5 mg Reaction
3 x 60 μg Reaction
5 x 1 mg Reaction
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-300-03 5 x 1 mg Reaction
70-300-04 1 x 5 mg Reaction
70-300-05 1 x 100 μg Reaction
70-300-15 1 mg Reaction
70-300-10 3 x 60 μg Reaction
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 70-300-03 Supplier Novus Biologicals™ Supplier No. 7030003
Add to Cart
Add to Cart

Applications: Flow Cytometry

R-Phycoerythrin (R-PE) is a fluorescent protein from the phycobiliprotein family, and is isolated from red algae. The absorbance spectrum of R-PE covers a broad range of excitation wavelengths, which provides an advantage for multi-color immunofluorescent staining or cell sorting. R-PE is one of the most intensely fluorescent phycobiliprotein having orange fluorescence. It is significantly brighter and more photostable than conventional organic fluorophores and it has a high quantum yield.

Order Info

Quantity

Specifications

Quantity 5 x 1 mg Reaction
Content And Storage Store at -20°C in the dark. Avoid freeze-thaw cycles.
Conjugate PE
Product Type R-PE Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.