Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Lightning-Link (R) R-PE Antibody Labeling Kit
SDP

Catalog No. 7030010 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg Reaction
1 x 100 μg Reaction
1 x 5 mg Reaction
3 x 60 μg Reaction
5 x 1 mg Reaction
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-300-10 3 x 60 μg Reaction
70-300-03 5 x 1 mg Reaction
70-300-04 1 x 5 mg Reaction
70-300-05 1 x 100 μg Reaction
70-300-15 1 mg Reaction
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 70-300-10 Supplier R&D Systems™ Supplier No. 7030010
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Applications: Flow Cytometry

  • Easy to use
  • Only requires 30 seconds of hands-on time
  • No spin or separation steps
  • Generates conjugates identical to or better than those prepared with laborious multistep conjugation procedures

Specifications

Product Type Antibody Labeling Kit
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Conjugate PE
Product Line Novus
Quantity 3 x 60 μg Reaction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.